ENA410542971 Enamine

ethyl 2-hydroxy-3-methoxypropanoate

  • MDL number MFCD21953469
  • Cas Number : 849736-99-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138695

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138695
Availability details for
Quantity
  • Availability
  • Add to Cart