ENA410542973 Enamine

5-bromo-1-(cyclopropylmethyl)-1,2-dihydropyridin-2-one

  • MDL number MFCD21746683
  • Cas Number : 1114573-41-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138713

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138713
Availability details for
Quantity
  • Availability
  • Add to Cart