ENA410542981 Enamine

3-chloro-5,6-dimethyl-1,2,4-triazine

  • MDL number MFCD18804545
  • Cas Number : 412923-53-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138779

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138779
Availability details for
Quantity
  • Availability
  • Add to Cart