ENA410542984 Enamine

N-(4-amino-3-methylphenyl)-3-chlorobenzamide

  • MDL number MFCD16508171
  • Cas Number : 1285097-46-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139...141
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138789

Safety Information

msds https://www.enaminestore.com/catalog/EN300-138789
Availability details for
Quantity
  • Availability
  • Add to Cart