ENA410542985 Enamine

3-(carbamoylmethoxy)-4-methylbenzoic acid

  • MDL number MFCD14647012
  • Cas Number : 1249590-78-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 243...245
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138790

Safety Information

msds https://www.enaminestore.com/catalog/EN300-138790
Availability details for
Quantity
  • Availability
  • Add to Cart