ENA410542990 Enamine

trimethylpyrazine-2-carboxylic acid

  • MDL number MFCD18377781
  • Cas Number : 186534-01-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138797

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138797
Availability details for
Quantity
  • Availability
  • Add to Cart