ENA410542995 Enamine

2-(4-fluorophenyl)-2-methylcyclopropan-1-amine hydrochloride

  • MDL number MFCD26407954
  • Cas Number : 1803605-97-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138802

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138802
Availability details for
Quantity
  • Availability
  • Add to Cart