ENA410542997 Enamine

2-chloro-5-(5-chloro-1,3-benzoxazol-2-yl)aniline

  • MDL number MFCD03031539
  • Cas Number : 443290-23-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 195...197
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138804

Safety Information

msds https://www.enaminestore.com/catalog/EN300-138804
Availability details for
Quantity
  • Availability
  • Add to Cart