ENA410542999 Enamine

1-{4-[(2-fluorophenyl)methoxy]phenyl}methanamine

  • MDL number MFCD09718028
  • Cas Number : 849807-01-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138806

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138806
Availability details for
Quantity
  • Availability
  • Add to Cart