ENA410543001 Enamine

2-[(4-aminophenyl)methyl]butanoic acid

  • MDL number MFCD20652338
  • Cas Number : 103893-69-2
  • Molecular Weight : 193.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(4-aminobenzyl)butanoic acid
Inchi Code 1S/C11H15NO2/c1-2-9(11(13)14)7-8-3-5-10(12)6-4-8/h3-6,9H,2,7,12H2,1H3,(H,13,14)
Inchi Key FUKUXANCTILXJK-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-138808

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138808
Availability details for
Quantity
  • Availability
  • Add to Cart