ENA410543020 Enamine

2-(2-bromophenyl)-1-(4-fluorophenyl)ethan-1-one

  • MDL number MFCD12605187
  • Cas Number : 1127955-69-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 66...68
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138903

Safety Information

msds https://www.enaminestore.com/catalog/EN300-138903
Availability details for
Quantity
  • Availability
  • Add to Cart