ENA410543029 Enamine

5-(bromomethyl)-3-(1-methyl-1H-pyrazol-4-yl)-1,2-oxazole

  • MDL number MFCD19665223
  • Cas Number : 1341608-81-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138922

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138922
Availability details for
Quantity
  • Availability
  • Add to Cart