ENA410543031 Enamine

ethyl 4-amino-3-phenyl-1,2-thiazole-5-carboxylate

  • MDL number MFCD00848440
  • Cas Number : 65237-13-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138925

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138925
Availability details for
Quantity
  • Availability
  • Add to Cart