ENA410543032 Enamine

11-oxatricyclo[6.2.1.0,2,7]undeca-2(7),3,5-trien-3-amine

  • MDL number MFCD20721468
  • Cas Number : 668492-12-4
  • Molecular Weight : 161.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 75-76
Iupac Name 1,2,3,4-tetrahydro-1,4-epoxynaphthalen-5-amine
Inchi Code 1S/C10H11NO/c11-7-3-1-2-6-8-4-5-9(12-8)10(6)7/h1-3,8-9H,4-5,11H2
Inchi Key JHILHWLHCSISQQ-UHFFFAOYSA-N

Description

Melting Point 75-76
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-138929

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-138929
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart