ENA410543041 Enamine

4-(aminomethyl)-N-(2-hydroxyphenyl)benzamide hydrochloride

  • MDL number MFCD26407967
  • Cas Number : 1607315-36-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138952

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138952
Availability details for
Quantity
  • Availability
  • Add to Cart