ENA410543043 Enamine

2-fluoro-2-phenylacetonitrile

  • MDL number MFCD26407968
  • Cas Number : 10036-43-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138972

Safety Information

msds https://www.enaminestore.com/catalog/EN300-138972
Availability details for
Quantity
  • Availability
  • Add to Cart