ENA410543053 Enamine

1-bromocyclohexane-1-carbonitrile

  • MDL number MFCD14702546
  • Cas Number : 676132-35-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139070

Safety Information

msds https://www.enaminestore.com/catalog/EN300-139070
Availability details for
Quantity
  • Availability
  • Add to Cart