ENA410543055 Enamine

1-(6-chloropyridin-2-yl)-4-methanesulfonylpiperazine

  • MDL number MFCD00139045
  • Cas Number : 260441-50-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110953

Safety Information

msds https://www.enaminestore.com/catalog/EN300-110953
Availability details for
Quantity
  • Availability
  • Add to Cart