ENA410543065 Enamine

2-(2-carbamoylpyrrolidin-1-yl)-2-oxoacetic acid

  • MDL number MFCD12171742
  • Cas Number : 1248930-14-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-139146

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139146
Availability details for
Quantity
  • Availability
  • Add to Cart