ENA410543067 Enamine

2-chloro-6-(cyclopentylsulfanyl)benzoic acid

  • MDL number MFCD18270809
  • Cas Number : 1338949-07-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 142...144
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139150

Safety Information

msds https://www.enaminestore.com/catalog/EN300-139150
Availability details for
Quantity
  • Availability
  • Add to Cart