ENA410543069 Enamine

1-(2-bromo-4-methoxyphenyl)-2-(3-fluorophenyl)ethan-1-one

  • MDL number MFCD12517678
  • Cas Number : 1184583-68-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139157

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139157
Availability details for
Quantity
  • Availability
  • Add to Cart