ENA410543072 Enamine

4-cyano-2-methoxybenzene-1-sulfonamide

  • MDL number MFCD18394204
  • Cas Number : 1261582-52-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139161

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139161
Availability details for
Quantity
  • Availability
  • Add to Cart