ENA410543075 Enamine

2-chloro-4,5-dimethylthiophene-3-carboxylic acid

  • MDL number MFCD18415682
  • Cas Number : 1261769-27-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139166

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139166
Availability details for
Quantity
  • Availability
  • Add to Cart