ENA410543077 Enamine

5-[(2-fluorophenyl)methoxy]-1,3-dimethyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD19395320
  • Cas Number : 1467884-48-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 191...193
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139168

Safety Information

msds https://www.enaminestore.com/catalog/EN300-139168
Availability details for
Quantity
  • Availability
  • Add to Cart