ENA410543089 Enamine

4-methyl-4,5-dihydro-1,3-oxazol-2-amine

  • MDL number MFCD19208609
  • Cas Number : 68210-17-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139263

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-139263
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart