ENA410543095 Enamine

(but-3-yn-1-yl)(2,2,2-trifluoroethyl)amine hydrochloride

  • MDL number MFCD26407993
  • Cas Number : 1607287-84-2
  • Molecular Weight : 187.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-(2,2,2-trifluoroethyl)but-3-yn-1-amine hydrochloride
Inchi Code 1S/C6H8F3N.ClH/c1-2-3-4-10-5-6(7,8)9;/h1,10H,3-5H2;1H
Inchi Key VKPNXTVXUAKPER-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-139279

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139279
Availability details for
Quantity
  • Availability
  • Add to Cart