ENA410543110 Enamine

methyl cyclohept-1-ene-1-carboxylate

  • MDL number MFCD26407515
  • Cas Number : 56745-53-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-111299

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111299
Availability details for
Quantity
  • Availability
  • Add to Cart