ENA410543117 Enamine

(cyclopent-1-en-1-yl)methanol

  • MDL number MFCD19646923
  • Cas Number : 1120-80-5
  • Molecular Weight : 98.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-cyclopenten-1-ylmethanol
Inchi Code 1S/C6H10O/c7-5-6-3-1-2-4-6/h3,7H,1-2,4-5H2
Inchi Key WJDFLCXFDSYKID-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-139379

Safety Information

signalword Warning
pcodes P210,P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P370+P378,P403+P233,P403+P235,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-139379
pictograms GHS07
hstatements H227,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart