ENA410543136 Enamine

7-chloro-1-methyl-1H-1,3-benzodiazol-2-amine

  • MDL number MFCD12111693
  • Cas Number : 945021-15-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 209...211
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139450

Safety Information

msds https://www.enaminestore.com/catalog/EN300-139450
Availability details for
Quantity
  • Availability
  • Add to Cart