ENA410543143 Enamine

1-methyl-4-nitro-1H-pyrazole-5-sulfonyl chloride

  • MDL number MFCD21647363
  • Cas Number : 1365941-03-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-139467
Availability details for
Quantity
  • Availability
  • Add to Cart