ENA410543146 Enamine

3-(2-carboxyethanesulfinyl)propanoic acid

  • MDL number MFCD02578993
  • Cas Number : 3680-08-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139473

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139473
Availability details for
Quantity
  • Availability
  • Add to Cart