ENA410543147 Enamine

3-fluoro-4-(oxolan-3-ylmethoxy)benzene-1-sulfonyl chloride

  • MDL number MFCD16204812
  • Cas Number : 1308327-46-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 75...77
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139482

Safety Information

msds https://www.enaminestore.com/catalog/EN300-139482
Availability details for
Quantity
  • Availability
  • Add to Cart