ENA410543155 Enamine

2-aminohex-5-enoic acid hydrochloride

  • MDL number MFCD26408012
  • Cas Number : 1237001-51-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 176...178
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139499

Safety Information

msds https://www.enaminestore.com/catalog/EN300-139499
Availability details for
Quantity
  • Availability
  • Add to Cart