ENA410543160 Enamine

2-(piperazin-1-yl)-4-(propan-2-yl)pyrimidine

  • MDL number MFCD18330617
  • Cas Number : 1342539-22-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139505

Safety Information

msds https://www.enaminestore.com/catalog/EN300-139505
Availability details for
Quantity
  • Availability
  • Add to Cart