ENA410543165 Enamine

N-(2-aminocyclohexyl)bicyclo[2.2.1]heptane-2-carboxamide hydrochloride

  • MDL number MFCD26408016
  • Cas Number : 1803598-79-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139549

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139549
Availability details for
Quantity
  • Availability
  • Add to Cart