ENA410543167 Enamine

(cyclopropylcarbamoyl)methyl 3,5-dimethoxybenzoate

  • MDL number MFCD02718365
  • Cas Number : 380193-95-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139551

Safety Information

msds https://www.enaminestore.com/catalog/EN300-139551
Availability details for
Quantity
  • Availability
  • Add to Cart