ENA410543172 Enamine

(6-phenyl-4,5,6,7-tetrahydro-1,3-benzothiazol-2-yl)methanamine

  • MDL number MFCD11129288
  • Cas Number : 1019351-57-5
  • Molecular Weight : 244.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139615

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139615
Availability details for
Quantity
  • Availability
  • Add to Cart