ENA410543175 Enamine

trimethylpyrimidine-5-carboxylic acid

  • MDL number MFCD11040459
  • Cas Number : 108397-75-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-139620

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139620
Availability details for
Quantity
  • Availability
  • Add to Cart