ENA410543177 Enamine

1-(4-aminopyridine-2-carbonyl)piperidin-4-ol

  • MDL number MFCD16719449
  • Cas Number : 1251365-98-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139623

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139623
Availability details for
Quantity
  • Availability
  • Add to Cart