ENA410543178 Enamine

1-(5-methylfuran-2-yl)piperazin-2-one

  • MDL number MFCD26408019
  • Cas Number : 1510374-30-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139624

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139624
Availability details for
Quantity
  • Availability
  • Add to Cart