ENA410543180 Enamine

2-(2-chlorophenyl)-2-methylpropanenitrile

  • MDL number MFCD11036574
  • Cas Number : 69849-08-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139626

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139626
Availability details for
Quantity
  • Availability
  • Add to Cart