ENA410543181 Enamine

2,3-dihydro-1-benzoxepine

  • MDL number MFCD22681566
  • Cas Number : 14949-49-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139627

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139627
Availability details for
Quantity
  • Availability
  • Add to Cart