ENA410543185 Enamine

ethyl 2-amino-3-(4-chloro-3-fluorophenyl)-3-hydroxypropanoate

  • MDL number MFCD22297102
  • Cas Number : 1498207-35-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139631

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139631
Availability details for
Quantity
  • Availability
  • Add to Cart