ENA410543188 Enamine

pyrazolo[1,5-b]pyridazine-3-carboxylic acid

  • MDL number MFCD11865261
  • Cas Number : 88561-91-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114593

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114593
Availability details for
Quantity
  • Availability
  • Add to Cart