ENA410543190 Enamine

2-(2-hydroxy-2-phenylethyl)cycloheptan-1-ol

  • MDL number MFCD22349316
  • Cas Number : 1559763-19-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139656

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139656
Availability details for
Quantity
  • Availability
  • Add to Cart