ENA410543191 Enamine

2-({[(tert-butoxy)carbonyl]amino}methyl)pyridine-4-carboxylic acid

  • MDL number MFCD11044225
  • Cas Number : 473924-63-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 189...191
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114708

Safety Information

msds https://www.enaminestore.com/catalog/EN300-114708
Availability details for
Quantity
  • Availability
  • Add to Cart