ENA410543197 Enamine

1-bromohex-3-ene

  • MDL number MFCD00153977
  • Cas Number : 84254-20-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139723

Safety Information

msds https://www.enaminestore.com/catalog/EN300-139723
Availability details for
Quantity
  • Availability
  • Add to Cart