ENA410543198 Enamine

1-methoxybut-3-yn-2-amine

  • MDL number MFCD21263569
  • Cas Number : 1481004-00-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139724

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139724
Availability details for
Quantity
  • Availability
  • Add to Cart