ENA410543201 Enamine

8-chloro-3-cyclopropyl-[1,2,4]triazolo[4,3-a]pyrazine

  • MDL number MFCD20303912
  • Cas Number : 140911-01-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 171...173
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139728

Safety Information

msds https://www.enaminestore.com/catalog/EN300-139728
Availability details for
Quantity
  • Availability
  • Add to Cart