ENA410543203 Enamine

tert-butyl (1R,5S,7s)-7-{[(benzyloxy)carbonyl]amino}-9-oxo-3-azabicyclo[3.3.1]nonane-3-carboxylate

  • MDL number MFCD26408026
  • Cas Number : 1638643-12-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139731

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139731
Availability details for
Quantity
  • Availability
  • Add to Cart